7-Aminoindole 95%

Supplier: Ambeed
Warning

Synonyms: Indol-7-ylamine, 1H-Indol-7-amine, 7-Amino-1H-indole, 7-Indolamine

A235435-100MG A235435-10G A235435-250MG A235435-1G A235435-5G
103811-014EA 37 USD
103811-014 103850-782 76771-250 76771-252 76771-254 77089-786 77089-788
7-Aminoindole 95%
7-Aminoindole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 354 °C (760 mm Hg)
Melting Pt: 96…100 °C
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD02093960
CAS Number: 5192-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR