6-Hydroxyindole 97%

Supplier: Ambeed
Danger

Synonyms: Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol

A135711-100MG A135711-250MG A135711-1G
103802-208EA 25 USD
103802-208 77272-234 77272-236 77315-054
6-Hydroxyindole 97%
6-Hydroxyindole

Precaución: for Research Use Only

Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 124…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00152101
CAS Number: 2380-86-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR