1H-Indazole-7-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A157120-100G
A157120-10G
A157120-250MG
A157120-1G
A157120-25G
A157120-5G
103804-968EA
5
USD
103804-968
103843-168
77163-010
77163-012
77163-014
77318-984
77318-986
77318-988
1H-Indazole-7-carboxylic acid 97%
1H-Indazole-7-carboxylic acid
Precaución: Sealed in dry, at room temperature.
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 443.7 °C (760 mm Hg) Flash Pt: 222.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD06804572 CAS Number: 677304-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...