1H-Indazole-6-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A187991-25G
A187991-100G
A187991-10G
A187991-5G
103816-062EA
6
USD
103816-062
103846-652
77084-370
77164-138
77164-140
77302-110
1H-Indazole-6-carboxylic acid 98%
1H-Indazole-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.1454 g/mol Boiling Pt: 443.7 °C (760 mm Hg) Melting Pt: 302…307 °C Storage Temperature: Ambient |
MDL Number:
MFCD06804571 CAS Number: 704-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...