1H-Indazole-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A119604-10G
A119604-250MG
A119604-25G
A119604-1G
A119604-5G
103799-800EA
7
USD
103799-800
77026-304
77026-306
77026-308
77297-702
77297-704
1H-Indazole-5-carboxylic acid 98%
1H-Indazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Melting Pt: 318…322 °C Storage Temperature: Ambient |
MDL Number:
MFCD06804570 CAS Number: 61700-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...