1H-Indazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A517750-1G A517750-25G A517750-5G A517750-250MG
103828-478EA 14 USD
103828-478 103839-582 76843-582 76843-584 76843-586 77169-486
1H-Indazole-5-carbaldehyde 97%
1H-Indazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.1500 g/mol
Boiling Pt: 358.3 °C (760 mm Hg)
Melting Pt: 115…119 °C
Flash Pt: 174.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06738280
CAS Number: 253801-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR