1H-Indazole-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A503101-1G
A503101-25G
A503101-5G
A503101-10G
A503101-250MG
103828-306EA
6
USD
103828-306
103839-400
76843-368
76843-370
76843-372
77169-346
77169-348
1H-Indazole-4-carboxylic acid 97%
1H-Indazole-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.15 g/mol Boiling Pt: 443.7 °C (760 mm Hg) Melting Pt: 288…293 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840644 CAS Number: 677306-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...