1H-Indazole-4-carbaldehyde 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A353752-1G
A353752-5G
A353752-250MG
A353752-100MG
103826-106EA
30
USD
103826-106
103837-108
76840-222
76840-224
77684-043
77892-724
1H-Indazole-4-carbaldehyde 98%, Ambeed.Inc
1H-Indazole-4-carbaldehyde
Precaución: For research use only.
Formula:
C₈H₆N₂O Boiling Pt: 358.3 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD06738279 CAS Number: 669050-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...