1H-Indazole-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Indazole-3-carbaldehyde

A290744-1G A290744-25G A290744-5G A290744-100G A290744-10G A290744-250MG
103813-874EA 27 USD
103813-874 103835-016 76837-596 76837-598 76837-600 77239-056 77239-058 77239-060
1H-Indazole-3-carboxaldehyde 98%
1H-Indazole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.146 g/mol
Boiling Pt: 193.6 °C (760 mm Hg)
Flash Pt: 71.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972468
CAS Number: 5235-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR