1H-Indazol-7-ol 97%

Supplier: Ambeed
Warning

Synonyms: 7-Hydroxy-1H-indazole

A133183-10G A133183-250MG A133183-25G A133183-1G A133183-5G
103801-838EA 55 USD
103801-838 76983-836 76983-838 76983-840 77314-460 77314-462
1H-Indazol-7-ol 97%
1H-Indazol-7-ol

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.1400 g/mol
Boiling Pt: 366.5 °C (760 mm Hg)
Flash Pt: 175.464 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00464298
CAS Number: 81382-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR