1H-Indazole-6-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Indazol-6-ylboronic acid, 6-Indazolyboronic acid, 1H-Indazol-6-yl-6-boronic acid

A172155-1G A172155-25G A172155-5G A172155-10G A172155-250MG
103806-700EA 12 USD
103806-700 103844-984 77082-164 77082-166 77082-168 77151-304 77151-306
1H-Indazole-6-boronic acid 97%
1H-Indazole-6-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
MW: 161.9500 g/mol
Melting Pt: 112…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972419
CAS Number: 885068-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR