3-Amino-1H-indazole 98%

Supplier: Ambeed
Warning

A135561-100G A135561-10G A135561-500G A135561-1G A135561-25G A135561-5G
103802-178EA 9 USD
103802-178 77234-690 77234-692 77234-694 77315-018 77315-020 77315-022
3-Amino-1H-indazole 98%
3-Amino-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
Boiling Pt: 376.6 °C (760 mm Hg)
Melting Pt: 156…157 °C
Flash Pt: 209.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00182045
CAS Number: 874-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR