1H-Imidazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: Imidazole-4-carboxylic acid

A183327-1G A183327-5G A183327-100G A183327-10G
103815-632EA 6 USD
103815-632 103846-194 77083-796 77083-798 77111-496 77111-498
1H-Imidazole-4-carboxylic acid 97%
1H-Imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 495 °C (760 mm Hg)
Melting Pt: 243…245 °C
Flash Pt: 253.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00082203
CAS Number: 1072-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR