1H-Imidazole-4-carbonitrile 98%

Supplier: Ambeed
Warning

A135745-100MG A135745-10G A135745-250MG A135745-1G A135745-25G A135745-5G
103802-212EA 8 USD
103802-212 77234-700 77234-702 77234-704 77315-060 77315-062 77315-064
1H-Imidazole-4-carbonitrile 98%
1H-Imidazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₃N₃
MW: 93.09 g/mol
Boiling Pt: 388.2 °C (760 mm Hg)
Flash Pt: 130.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233358
CAS Number: 57090-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR