Imidazole hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Imidazole hydrochloride

A213325-100G A213325-10G A213325-500G A213325-5G A213325-25G A213325-1000G
103848-998EA 8 USD
103848-998 76800-690 76800-692 76800-694 76800-696 77087-506 77667-788
Imidazole hydrochloride 95%
Imidazole hydrochloride

Precaución: for Research Use Only

Formula: C₃H₄N₂·ClH
MW: 104.538 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00012695
CAS Number: 1467-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR