2-Aminoimidazole hemisulfate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)
A209404-100G
A209404-10G
A209404-500G
A209404-1G
A209404-25G
A209404-5G
103817-940EA
5
USD
103817-940
103848-626
76825-842
76825-844
76825-846
77086-970
77086-972
77086-974
2-Aminoimidazole hemisulfate 97%
2-Aminoimidazole hemisulphate
Precaución: for Research Use Only
Formula:
C₆H₁₂N₆O₄S MW: 264.26 g/mol Melting Pt: 270 °C Flash Pt: 168.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013162 CAS Number: 1450-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...