1H-Imidazol-2-amine 97%

Supplier: Ambeed
Warning

A184764-100MG A184764-250MG A184764-1G A184764-25G
103815-780EA 35 USD
103815-780 103846-346 76872-870 76872-872 77083-966 77671-616
1H-Imidazol-2-amine 97%
1H-Imidazol-2-amine

Precaución: for Research Use Only

Formula: C₃H₅N₃
Boiling Pt: 313.1 °C (760 mm Hg)
Flash Pt: 168.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00792530
CAS Number: 7720-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR