1H-Imidazo[4,5-b]pyridine-2-thiol 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Imidazo[4,5-b]pyridine-2(3H)-thione, 2-Thio-1H-imidazo[4,5-b]pyridine

A140900-100MG A140900-250MG A140900-25G A140900-1G A140900-5G
103802-918EA 33 USD
103802-918 77234-938 77234-940 77234-942 77316-036 77316-038
1H-Imidazo[4,5-b]pyridine-2-thiol 95%
1H-Imidazo[4,5-b]pyridine-2-thiol

Precaución: for Research Use Only

Formula: C₆H₅N₃S
MW: 151.189 g/mol
Boiling Pt: 308.3 °C (760 mm Hg)
Flash Pt: 140.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00022663
CAS Number: 29448-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR