1H-Imidazo[4,5-b]pyridin-2(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydro-2H-imidazo[4,5-b]pyridin-2-one

A100981-250MG A100981-1G A100981-5G A100981-10G
103795-732EA 26 USD
103795-732 103821-424 77188-538 77282-628 77296-932 77745-770
1H-Imidazo[4,5-b]pyridin-2(3H)-one 97%
1H-Imidazo[4,5-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₆H₅N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00464845
CAS Number: 16328-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR