1-Hexyl-3-methylimidazolium hexafluorophosphate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)

A130531-100G A130531-500G A130531-25G A130531-5G A130531-10G
77027-044EA 72 USD
77027-044 77027-046 77298-410 77313-874 77666-872
1-Hexyl-3-methylimidazolium hexafluorophosphate 95%
1-Hexyl-3-methylimidazolium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₉F₆N₂P
MW: 312.24
Melting Pt: 73.5 °C
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD03093296
CAS Number: 304680-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR