1H-Benzimidazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A145671-100G A145671-10G A145671-250MG A145671-25G A145671-1G A145671-5G A145671-500G
103803-568EA 9 USD
103803-568 103841-650 76997-064 76997-066 76997-068 76997-070 77299-468 77316-958 77671-068
1H-Benzimidazole-5-carboxylic acid 97%
1H-Benzimidazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011555
CAS Number: 15788-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR