Benzimidazole-5,6-dicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d]imidazole-5,6-dicarboxylic acid

A155696-100MG A155696-10G A155696-250MG A155696-1G A155696-5G
103804-790EA 15 USD
103804-790 103842-992 77028-356 77028-358 77028-360 77318-716 77318-718
Benzimidazole-5,6-dicarboxylic acid 97%
Benzimidazole-5,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆N₂O₄
MW: 206.16 g/mol
Boiling Pt: 660.3 °C (760 mm Hg)
Flash Pt: 353.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03093058
CAS Number: 10351-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR