1H-Benzo[d]imidazole-5,6-diamine dihydrochloride 97%

Supplier: Ambeed
Warning

A240224-100MG A240224-10G A240224-250MG A240224-1G A240224-5G
103811-342EA 125 USD
103811-342 103851-140 76987-610 76987-612 76987-614 77090-292 77090-294
1H-Benzo[d]imidazole-5,6-diamine dihydrochloride 97%
1H-Benzo[d]imidazole-5,6-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀Cl₂N₄
MW: 221.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12024649
CAS Number: 90000-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR