1H-Benzimidazole-2-carbaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d]imidazole-2-carbaldehyde

A127912-100MG A127912-250MG
103801-038EA 28 USD
103801-038 77234-368 77234-370
1H-Benzimidazole-2-carbaldehyde 95%
1H-Benzimidazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
Boiling Pt: 361.7 °C (760 mm Hg)
Melting Pt: 224 °C
Flash Pt: 176.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00228045
CAS Number: 3314-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR