4-Hydroxybenzimidazole 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-benzo[d]imidazol-4-ol, 1H-Benzimidazol-4-ol
A143921-100MG
A143921-250MG
A143921-1G
103803-346EA
50
USD
103803-346
77235-130
77235-132
77316-650
4-Hydroxybenzimidazole 95%
4-Hydroxybenzimidazole
Precaución: for Research Use Only
Formula:
C₇H₆N₂O MW: 134.14 g/mol Boiling Pt: 469.1 °C (760 mm Hg) Flash Pt: 237.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00963978 CAS Number: 67021-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...