5-Benzotriazolecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Benzo[d][1,2,3]triazole-5-carboxylic acid, 5-Carboxybenzotriazole

A194808-25G A194808-100G A194808-10G A194808-500G A194808-5G
103847-306EA 7 USD
103847-306 77085-234 77139-206 77139-208 77139-210 77302-436
5-Benzotriazolecarboxylic acid 97%
5-Benzotriazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Boiling Pt: 455.9 °C (760 mm Hg)
Melting Pt: 305 °C
Storage Temperature: Ambient
MDL Number: MFCD00012318
CAS Number: 23814-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR