1-Aminobenzotriazole 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Benzo[d][1,2,3]triazol-1-amine
A336445-1G
A336445-50MG
A336445-5G
A336445-100MG
A336445-1MG
A336445-250MG
A336445-25G
103799-608EA
6
USD
103799-608
103825-718
103836-692
76839-696
76839-698
76839-700
77195-086
77195-088
77195-090
77195-092
1-Aminobenzotriazole 95%
1-Aminobenzotriazole
Precaución: for Research Use Only
Formula:
C₆H₆N₄ MW: 134.14 g/mol Boiling Pt: 319.3 °C (760 mm Hg) Melting Pt: 81…84 °C Flash Pt: 146.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132902 CAS Number: 1614-12-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...