1H-Benzimidazole-7-carboxylic acid 98%

Supplier: Ambeed
Warning

A257301-1G A257301-250MG
103812-292EA 72 USD
103812-292 103852-160 77091-668 77113-578
1H-Benzimidazole-7-carboxylic acid 98%
1H-Benzimidazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Boiling Pt: 542.5 °C (760 mm Hg)
Melting Pt: 326…330 °C
Flash Pt: 281.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11656261
CAS Number: 46006-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR