1H-Benzimidazole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A285381-1G A285381-5G A285381-100G
103813-674EA 13 USD
103813-674 103834-808 76837-344 76837-346 77166-684 77166-686
1H-Benzimidazole-2-carboxylic acid 97%
1H-Benzimidazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
Boiling Pt: 334.5 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 222.318 °C
Storage Temperature: Freezer
MDL Number: MFCD00721565
CAS Number: 2849-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR