1,2,3-Triazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-1H-Triazole, 1H-1,2,3-Triazole

A242543-25G A242543-5G A242543-100G A242543-10G A242543-1G A242543-500G
103851-286EA 6 USD
103851-286 77090-462 77090-464 77165-792 77165-794 77165-796 77165-798
1,2,3-Triazole 98%
1,2,3-Triazole

Precaución: for Research Use Only

Formula: C₂H₃N₃
Boiling Pt: 203 °C (760 mm Hg)
Melting Pt: 23…25 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00014490
CAS Number: 288-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR