1H-1,2,3-Triazolo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 1H-[1,2,3]Triazolo[4,5-b]pyridine, 4-Azabenzotriazole

A208259-1G A208259-10G A208259-250MG
103817-866EA 44 USD
103817-866 103848-554 77086-862 77237-318 77567-998
1H-1,2,3-Triazolo[4,5-b]pyridine 95%
1H-1,2,3-Triazolo[4,5-b]pyridine

Precaución: For research use only.

Formula: C₅H₄N₄
Boiling Pt: 204.5 °C (760 mm Hg)
Flash Pt: 68.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00134183
CAS Number: 273-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR