1H-[1,2,3]Triazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1H-[1,2,3]Triazole-4-carboxylic acid, 3H-[1,2,3]Triazole-4-carboxylic acid, 1H-1,2,3-Triazole-4-carboxylic acid, 3H-1-2-3-Triazole-4-carboxylic-acid, 4-Carboxy-1H-1,2,3-triazole

A163811-250MG A163811-1G A163811-5G
103805-770EA 23 USD
103805-770 103844-034 77191-190 77320-148 77320-150
1H-[1,2,3]Triazole-4-carboxylic acid 98%
1H-[1,2,3]Triazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₃H₃N₃O₂
Boiling Pt: 446.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00196046
CAS Number: 16681-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR