2,2'-Biimidazole 97%

Supplier: Ambeed
Warning

A131787-250MG A131787-1G A131787-25G
103801-630EA 9 USD
103801-630 77246-560 77314-154 77314-156
2,2'-Biimidazole 97%
2,2'-Biimidazole

Precaución: for Research Use Only

Formula: C₆H₆N₄
Flash Pt: 238.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00047014
CAS Number: 492-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR