1-Fluorocyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

A103096-10G A103096-250MG A103096-1G A103096-5G
103796-480EA 15 USD
103796-480 103822-184 76994-634 76994-636 77309-056 77309-058
1-Fluorocyclopropanecarboxylic acid 97%
1-Fluorocyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅FO₂
MW: 104.08 g/mol
Melting Pt: 63…67 °C
Flash Pt: 56.6 °C
Storage Temperature: Ambient
MDL Number: MFCD18205954
CAS Number: 137081-41-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR