4-Fluoro-3-nitroanisole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Fluoro-4-methoxy-2-nitrobenzene
A446405-100G
A446405-10G
A446405-250MG
A446405-1G
A446405-25G
A446405-5G
103827-744EA
6
USD
103827-744
103838-810
76789-310
76789-312
76789-314
76842-608
76842-610
76842-612
4-Fluoro-3-nitroanisole 98%
4-Fluoro-3-nitroanisole
Precaución: for Research Use Only
Formula:
C₇H₆FNO₃ Boiling Pt: 279.5 °C (760 mm Hg) Flash Pt: 122.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD11848442 CAS Number: 61324-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...