2-Fluoro-5-nitroanisole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Fluoro-2-methoxy-4-nitrobenzene, 4-Fluoro-3-methoxynitrobenzene
A199656-25G
A199656-5G
A199656-100G
A199656-10G
A199656-500G
A199656-1G
103817-120EA
9
USD
103817-120
103847-746
77085-790
77085-792
77260-760
77260-762
77260-764
77302-622
2-Fluoro-5-nitroanisole 97%
2-Fluoro-5-nitroanisole
Precaución: for Research Use Only
Formula:
C₇H₆FNO₃ Boiling Pt: 270.3±20.0 °C (760 mm Hg) Melting Pt: 69…71 °C Flash Pt: 117.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782081 CAS Number: 454-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...