1-Fluoro-2-iodobenzene 98% stabilized

Supplier: Ambeed
Danger

Synonyms: 2-Fluoroiodobenzene

Stabilization: Stabilized with Copper chip 0.01%

A505396-25G A505396-5G A505396-100G
103839-424EA 14 USD
103839-424 76843-400 76843-402 77617-160
1-Fluoro-2-iodobenzene 98% stabilized
1-Fluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 189.3 °C (760 mm Hg)
Flash Pt: 71.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001031
CAS Number: 348-52-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR