1-Ethynyl-4-nitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenylacetylene

A123031-10G A123031-25G A123031-1G A123031-5G
103800-332EA 6 USD
103800-332 77234-198 77234-200 77234-202 77312-448 77312-450
1-Ethynyl-4-nitrobenzene 97%
1-Ethynyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₅NO₂
Boiling Pt: 246.6 °C (760 mm Hg)
Melting Pt: 148…150 °C
Flash Pt: 112.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00024794
CAS Number: 937-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR