1-Ethynyl-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Trifluoromethoxy)phenylacetylene

A749576-250MG A749576-1G
103840-846EA 56 USD
103840-846 76791-324 76845-078
1-Ethynyl-4-(trifluoromethoxy)benzene 98%
1-Ethynyl-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₉H₅F₃O
MW: 186.13 g/mol
Boiling Pt: 158.1 °C (760 mm Hg)
Flash Pt: 40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094333
CAS Number: 160542-02-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR