2-Nitrophenylacetylene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethynyl-2-nitrobenzene

A190091-5G A190091-100MG A190091-250MG
103816-258EA 29 USD
103816-258 103846-854 77084-644 77236-852 77236-854
2-Nitrophenylacetylene 97%
2-Nitrophenylacetylene

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MW: 147.131 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01113917
CAS Number: 16433-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR