4-Ethylphenylacetylene 98%

Supplier: Ambeed
Warning

Synonyms: (4-Ethylphenyl)acetylene, 1-Ethyl-4-ethynylbenzene, 1-Ethynyl-4-ethylbenzene, (4-Ethylphenyl)ethyne

A160173-100G A160173-10G A160173-500G A160173-1G A160173-25G A160173-5G
103805-344EA 5 USD
103805-344 103843-574 76689-796 76689-798 76689-800 77300-446 77319-494 77319-496
4-Ethylphenylacetylene 98%
4-Ethylphenylacetylene

Precaución: for Research Use Only

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Boiling Pt: 183.7 °C (760 mm Hg)
Flash Pt: 71.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00173887
CAS Number: 40307-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR