1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Ethyl-1H-pyrazol-4yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Ethyl-1H-pyrazole-4-boronic acid Pinacol ester, 1-Ethyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A166804-10G A166804-250MG A166804-1G A166804-25G A166804-5G
103806-100EA 14 USD
103806-100 103844-362 76998-044 76998-046 77300-848 77300-850 77300-852
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester 95%
1-Ethyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
MW: 222.0918 g/mol
Boiling Pt: 322.6 °C (760 mm Hg)
Melting Pt: 44…49 °C
Flash Pt: 148.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864188
CAS Number: 847818-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR