1-Ethyl-3-methylimidazolium acetate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-methylimidazolium Acetate

A214853-100G A214853-10G A214853-500G A214853-1G A214853-25G A214853-5G
103818-438EA 6 USD
103818-438 103849-140 77152-576 77152-578 77152-580 77303-380 77303-382 77303-384
1-Ethyl-3-methylimidazolium acetate 95%
1-Ethyl-3-methylimidazolium acetate

Precaución: for Research Use Only

Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
Melting Pt: …20 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD06798186
CAS Number: 143314-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR