1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethyl-3-methylpyrazole-5-carboxylic acid

A139361-100G A139361-10G A139361-1G A139361-25G A139361-5G A139361-500G
103802-734EA 17 USD
103802-734 77109-748 77109-750 77315-820 77315-822 77315-824 77685-192
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid 97%
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
Boiling Pt: 308.5 °C (760 mm Hg)
Melting Pt: 132…145 °C
Flash Pt: 140.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00156147
CAS Number: 50920-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR