1-Ethyl-3-methylimidazoliumhexafluorophosphate 97%
Supplier: Ambeed
|
Warning
|
A290131-100G
A290131-5G
103835-002EA
52
USD
103835-002
77239-044
77305-574
1-Ethyl-3-methylimidazoliumhexafluorophosphate 97%
1-Ethyl-3-methylimidazoliumhexafluorophosphate
Precaución: for Research Use Only
Formula:
C₆H₁₁F₆N₂P Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216703 CAS Number: 155371-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...