1-Ethyl-1H-1,2,3-triazole 97%

Supplier: Ambeed
Warning

A612314-100MG A612314-10G A612314-1G A612314-25G A612314-5G
103840-172EA 166 USD
103840-172 76650-202 76650-204 76844-338 76844-340 76844-342
1-Ethyl-1H-1,2,3-triazole 97%
1-Ethyl-1H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₄H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD18324607
CAS Number: 78910-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR