1-Ethyl-1H-1,2,4-triazole 97%

Supplier: Ambeed
Warning

A101352-250MG A101352-1G A101352-5G
103795-924EA 33 USD
103795-924 103821-612 77233-076 77297-064 77297-066
1-Ethyl-1H-1,2,4-triazole 97%
1-Ethyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₄H₇N₃
Boiling Pt: 188.1 °C (760 mm Hg)
Flash Pt: 67.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07367971
CAS Number: 16778-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR