1-Decyl-1H-imidazole 95%

Supplier: Ambeed
Warning

A185527-250MG A185527-25G A185527-5G
103815-864EA 14 USD
103815-864 103846-432 77164-036 77164-038 77301-982
1-Decyl-1H-imidazole 95%
1-Decyl-1H-imidazole

Precaución: Sealed in dry, at 2…8°C

Formula: C₁₃H₂₄N₂
MW: 208.34 g/mol
Boiling Pt: 333.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD19011534
CAS Number: 33529-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR