1-Cyclopropyl-8-ethoxy-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A115482-250MG A115482-1G
103798-570EA 519 USD
103798-570 76982-940 77297-372
1-Cyclopropyl-8-ethoxy-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 97%
1-Cyclopropyl-8-ethoxy-6-fluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₄FN₃O₄
MW: 389.42 g/mol
Boiling Pt: 613.4 °C (760 mm Hg)
Flash Pt: 324.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26743466
CAS Number: 182868-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR