1-Cyclopropyl-2-(2-fluorophenyl)ethanone 95%
Supplier: Ambeed
|
Warning
|
A150399-100G
A150399-25G
A150399-500G
A150399-5G
A150399-1G
A150399-10G
103804-158EA
5
USD
103804-158
103842-292
77095-796
77095-798
77095-800
77299-800
77317-808
77685-251
1-Cyclopropyl-2-(2-fluorophenyl)ethanone 95%
1-Cyclopropyl-2-(2-fluorophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₁₁H₁₁FO Boiling Pt: 253.6 °C (760 mm Hg) Flash Pt: 107.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD10687164 CAS Number: 150322-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...